Order Entry
Denmark
ContactUsLinkComponent
 
Human Recombinant TGF-B 1 (from CHO cells)
 
undefined
Human Recombinant TGF-B 1 (from CHO cells)

 

  • Conjugation:
    Unconjugated
  • Protein function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    −20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Biological activity:
    Bioactivity assay: Inhibition of IL-4-induced HT-2 cell proliferation, ED50 ≤0,5 ng/ml, bioactivity ≥2,0×10⁶ units/mg
  • Gene ID:
    P01137
  • Reconstitution instructions:
    Sterile 10 mM HCl at 0,1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    N
  • Protein synonyms:
    cartilage-inducing factor|Differentiation inhibiting factor
  • UniProtKB:
    P01137
  • Protein/peptide name:
    TGF-beta1
  • Purity:
    ≥95%
  • Molecular weight:
    12,8/25,6 kDa
  • Sequence:
    ALDTNYCFSS TEKNCCVRQL YIDFRKDLGW KWIHEPKGYH ANFCLGPCPY IWSLDTQYSK VLALYNQHNP GASAAPCCVP QALEPLPIVY YVGRKPKVEQ LSNMIVRSCK CS
  • Endotoxin level:
    Low
  • Formulation:
    0,1% trifluoroacetic acid (TFA) and trehalose in a 20:1 trehalose to protein ratio
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT
  • Tested applications:
    Bioassay

 

Frequently Bought Together