Sodium L(+)-ascorbate
Glass bottle for solids, 100 g
Catalog # SERA14033.01
Marketsource
Supplier: Serva
Jump to:
Product Details & Documents
Synonyms: Sodium ascorbate, L(+)-Ascorbic acid sodium salt, L-Ascorbic acid sodium salt, Sodium (R)-2-((S)-1,2-dihydroxyethyl)-4-hydroxy-5-oxo-2,5-dihydrofuran-3-olate, Acidum ascorbicum sodium salt
- CAS Number:134-03-2
- Formula:C₆H₇NaO₆
- MDL Number:MFCD00082340
- Molecular Weight:198.11 g/mol
- Melting Pt:219 °C
- Density:1.799 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:205-126-1
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



