Sodium L(+)-ascorbate ≥99%, TECHNICAL
Supplier: VWR Chemicals
About this item
Synonyms: Sodium ascorbate, L(+)-Ascorbic acid sodium salt, L-Ascorbic acid sodium salt, Sodium (R)-2-((S)-1,2-dihydroxyethyl)-4-hydroxy-5-oxo-2,5-dihydrofuran-3-olate, Acidum ascorbicum sodium salt
- CAS Number:134-03-2
- Formula:C₆H₇NaO₆
- MDL Number:MFCD00082340
- Molecular Weight:198.11 g/mol
- Melting Pt:219 °C
- Density:1.799 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:205-126-1
2 Options Available Below
Product Details & Documents
Synonyms: Sodium ascorbate, L(+)-Ascorbic acid sodium salt, L-Ascorbic acid sodium salt, Sodium (R)-2-((S)-1,2-dihydroxyethyl)-4-hydroxy-5-oxo-2,5-dihydrofuran-3-olate, Acidum ascorbicum sodium salt
- CAS Number:134-03-2
- Formula:C₆H₇NaO₆
- MDL Number:MFCD00082340
- Molecular Weight:198.11 g/mol
- Melting Pt:219 °C
- Density:1.799 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:205-126-1
Specifications for Product Family
Specification Test Results
- Assay (on anhydrous substance):Min. 99 %
Product Family Options
Product Information
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



