Sodium L(+)-ascorbate ≥99%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 99% CAS No.: 134-03-2 Molecular Formula: C6H7NaO6 Formula Weight: 198.11 Melting Point: ca 220° Beilstein Reference: 3767246 Merck Reference: 14,830 Optical Rotation: +105° (c=5 in water) MDL No.: MFCD00082340
Synonyms: Sodium ascorbate, L(+)-Ascorbic acid sodium salt, L-Ascorbic acid sodium salt, Sodium (R)-2-((S)-1,2-dihydroxyethyl)-4-hydroxy-5-oxo-2,5-dihydrofuran-3-olate, Acidum ascorbicum sodium salt
- CAS Number:134-03-2
- Formula:C₆H₇NaO₆
- MDL Number:MFCD00082340
- Molecular Weight:198.11 g/mol
- Merck Index:14,830
- Melting Pt:∼220 °C
- Density:1.799 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:205-126-1
2 Options Available Below
Product Details & Documents
Purity: 99% CAS No.: 134-03-2 Molecular Formula: C6H7NaO6 Formula Weight: 198.11 Melting Point: ca 220° Beilstein Reference: 3767246 Merck Reference: 14,830 Optical Rotation: +105° (c=5 in water) MDL No.: MFCD00082340
Synonyms: Sodium ascorbate, L(+)-Ascorbic acid sodium salt, L-Ascorbic acid sodium salt, Sodium (R)-2-((S)-1,2-dihydroxyethyl)-4-hydroxy-5-oxo-2,5-dihydrofuran-3-olate, Acidum ascorbicum sodium salt
- CAS Number:134-03-2
- Formula:C₆H₇NaO₆
- MDL Number:MFCD00082340
- Molecular Weight:198.11 g/mol
- Merck Index:14,830
- Melting Pt:∼220 °C
- Density:1.799 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:205-126-1
Documents
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



