Sodium L(+)-ascorbate 99%
Supplier: Thermo Fisher Scientific
Om denne vare
Synonymer: Sodium ascorbate, L(+)-Ascorbic acid sodium salt, L-Ascorbic acid sodium salt, Sodium (R)-2-((S)-1,2-dihydroxyethyl)-4-hydroxy-5-oxo-2,5-dihydrofuran-3-olate, Acidum ascorbicum sodium salt
- CAS nummer:134-03-2
- Formel:C₆H₇NaO₆
- MDL nummer:MFCD00082340
- Molecular Weight:198.11 g/mol
- Smeltende Pt:219…221 °C (dec.)
- Tæthed:1,799 g/cm³ (20 °C)
- Opbevaringstemperatur:Ambient
- EINECS:205-126-1
4 tilgængelige muligheder nedenfor
Product Details & Documents
Synonymer: Sodium ascorbate, L(+)-Ascorbic acid sodium salt, L-Ascorbic acid sodium salt, Sodium (R)-2-((S)-1,2-dihydroxyethyl)-4-hydroxy-5-oxo-2,5-dihydrofuran-3-olate, Acidum ascorbicum sodium salt
- CAS nummer:134-03-2
- Formel:C₆H₇NaO₆
- MDL nummer:MFCD00082340
- Molecular Weight:198.11 g/mol
- Smeltende Pt:219…221 °C (dec.)
- Tæthed:1,799 g/cm³ (20 °C)
- Opbevaringstemperatur:Ambient
- EINECS:205-126-1
Documents
Specifications for Product Family
Specifikationstestresultater
- Appearance:White to yellow Crystalline powder
- Infrared spectrum:Conforms
- Titration Iodimetric:98.5 to 101.5 %
- Solubility:(5% in water) Clear colorless to light yellow
- Specific optical rotation:+103° to +108° (20°C, 589 nm) (c=5, H2O)
Product Family Options
Produktinformation
- Pack typePkAvailabilityPrice
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



