Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:(20 °C) insoluble
- Melting Point:53 °C
- Molar Mass:963.55 g/mol
- Boiling Point:100 °C (1013 hPa)
- Vapor Pressure:- 1.4 hPa (20 °C)
- Flash Point:149 °C
- Density:0.98 g/cm3 (25 °C)
Specification Test Results
Related Information
Frequently Bought Together