Span® 80, Span® for synthesis, Sigma-Aldrich®
: Merck
: Arlacel 80, Sorbitan monooleate
:1338-43-8
:
:C₂₄H₄₄O₆
:MFCD00080948
:1 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
:428.61 g/mol
:Ambient
:215-665-4
- Environmentally Preferable:
- Solubility:(20 °C) insoluble
- Molar Mass:428.60 g/mol
- Boiling Point:100 °C
- Flash Point:- 1.4 hPa (20 °C)
- Refractive Index:149 °C
- Density:1.0 g/cm3 (20 °C)
- Synonyms:Sorbitan monooleate, (Z)-Sorbitan mono-9-octadecenoate
- Hill Formula:C24H44O6
Frequently Bought Together