Order Entry
Denmark
ContactUsLinkComponent
 
 
 
Manganese(IV) oxide for synthesis
 :  Merck
 
 
 
Manganese(IV) oxide for synthesis
 :  Merck
 

 

  • Solubility:
    (20 °C) insoluble
  • Molar Mass:
    86.94 g/mol
  • Bulk Density:
    600 - 800 kg/m3
  • Vapor Pressure:
    (20 °C)
  • Density:
    5.03 g/cm3 (20 °C)
  • pH:
    (H2O) not applicable

 

Frequently Bought Together