Manganese(II) bromide, anhydrous 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13446-03-2
:
Formula:Br₂Mn
MDL Number:MFCD00011113
:214.75 g/mol
Merck Index:12,05765
Melting Pt:698 °C
Density:4.38 g/cm³
EINECS:236-591-9
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Pink Powder
- Titration Complexometric:≥98.5 %
Specification Test Results
Frequently Bought Together