Order Entry
Denmark
ContactUsLinkComponent
Titanium(IV) chloride for synthesis, Sigma-Aldrich®
Toxic
Corrosive
Titanium(IV) chloride for synthesis, Sigma-Aldrich®
Catalog # 8.12382.1000
Supplier:  Merck
CAS Number:  
Toxic
Corrosive
Titanium(IV) chloride for synthesis, Sigma-Aldrich®
Catalog # 8.12382.1000
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, plastic-coated
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :189.69 g/mol
Merck Index:13,09555
Boiling Pt:136 °C (1013 hPa)
Melting Pt:136.2 °C
Density:1.72 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-441-9
UN:1838
ADR:6.1 (8),I
 :M-Clarity™ quality level = MQ200
 :GHS06
 :GHS05
CAS Number:7550-45-0
Formula:Cl₄Ti
MDL Number:MFCD00011267