Bottles, wide neck, LDPE
: RIXIUS
LDPE, natural, with screw caps.
- Resistant to acids, alkalis and other chemicals
- Temperature resistant: –20 to +70 °C
Perfect for sample collection, storage and transport. Ideal for paste-like media and granular material. The wide mouth allows easy filling and cleaning.
: *Produktet/produkterne på denne side er uden for anvendelsesområdet for EU -direktiv 2019/904
Frequently Bought Together