Order Entry
Denmark
ContactUsLinkComponent
Cadmium(II) bromide ≥99.999% (metals basis)

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Cadmium bromide
CAS Number:7789-42-6
 :
Formula:Br₂Cd
MDL Number:MFCD00150204
 :272.22 g/mol
Merck Index:12,01651
Boiling Pt:863 °C (1013 hPa)
Melting Pt:567 °C
Density:5.19 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:232-165-1
UN:2570
ADR:6.1,III
 :GHS07
 :GHS09
MDL Number:MFCD00015996