Supelco® Organic Reference Standard, Acenaphthene
48500-U
: Merck
:
- Pk:5.000 mg
- Standards:Analytical standard
- CAS No.:83-32-9
- Description:Acenaphthene
- Storage conditions:2...30 °C
- Hazard class description:H410
- Shipping conditions:STD
- MDL No.:MFCD00003807
- M-Clarity quality level:MQ100
- Synonym(s):1,8-Ethylenenaphthalene
- Melting point:90 - 94 °C (lit.)
- Molecular weight:154,21
- Flash point:125 °C
- Boiling point:279 °C (lit.)
- GHS signal: GHS07, GHS09
: P273 - P391 - P501