Order Entry
Denmark
ContactUsLinkComponent
 
 
1,3-Dihydroxynaphthalene ≥99.00%, crystalline, Sigma-Aldrich®
 
undefined
 
1,3-Dihydroxynaphthalene ≥99.00%, crystalline, Sigma-Aldrich®

 

  • Appearance (colour):
    White to brown
  • Appearance (form):
    Powder
  • Purity (HPLC area %):
    ≥ 99 %
  • Solubility (colour):
    Light brown-yellow to very dark brown-yellow to very dark brown
  • Solubility (turbidity):
    Clear to slightly hazy
  • Solubility (method):
    50mg/ml ethanol
  • Recommended retest period:
    48 months

 

Frequently Bought Together