Neodyn(III)oxid, Sigma-Aldrich®
1.12276.0025
: Merck
:
- Pakning:Glass bottle
- Pakn:25 g
- Seq:3308
- Solubility:(20 °C) practically insoluble
- Melting Point:2272 °C
- Molar Mass:336.48 g/mol
- Bulk Density:1200 kg/m3
- Density:7.24 g/cm3 (20 °C)
:
:336.48 g/mol
:14,06450
:3760 °C (1013 hPa)
:2270 °C
:7,24 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
:215-214-1
:1313-97-9
:Nd₂O₃
:MFCD00011134