Cyanogen bromide for synthesis, Sigma-Aldrich®
Catalog # 8.20193.0050
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:50 g
- Seq:3368
- Environmentally Preferable:
Specifications
About this item
:
:105.92 g/mol
Merck Index:12,02763
Boiling Pt:61.4 °C (1013 hPa)
Melting Pt:52 °C
Density:2.06 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:208-051-2
UN:1889
ADR:6.1 (8),I
:M-Clarity™ quality level = MQ200
:GHS06
:GHS05
:GHS09
CAS Number:506-68-3
Formula:CBrN
MDL Number:MFCD00011597