Order Entry
Denmark
ContactUsLinkComponent
Cyanogen bromide 97%
Cyanogen bromide 97%
Certificates
undefined
Cyanogen bromide 97%

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    White Crystals
  • Titration Argentometric:
    96.0 to 103.0 %
  • Identification:
    Bromides : positive
  • Clarity of solution:
    (Aqueous) Passes test
  • Clarity of alcohol solution:
    Passes test

Specification Test Results

Related Information

Frequently Bought Together