Order Entry
Denmark
ContactUsLinkComponent
Ammonium peroxodisulphate (APS) ≥97.5%, GPR RECTAPUR®
Oxidizing
Irritant
Health
Ammonium peroxodisulphate (APS) ≥97.5%, GPR RECTAPUR®
Catalog # 21299.366
Supplier:  VWR Chemicals
CAS Number:  
undefined
Oxidizing
Irritant
Health
Ammonium peroxodisulphate (APS) ≥97.5%, GPR RECTAPUR®
Catalog # 21299.366
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    5 kg
  • Seq:
    4114

Specifications

Specification Test Results

  • Assay:
    Min. 97.5 %
  • Heavy metals (as Pb):
    Max. 50 ppm
  • Cl (Chloride):
    Max. 20 ppm
  • Fe (Iron):
    Max. 50 ppm

Specification Test Results

About this item

 :
 :228.2 g/mol
Merck Index:13,00545
Melting Pt:120 °C
Density:1.98 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-786-5
UN:1444
ADR:5.1,III
 :GHS03
 :GHS07
 :GHS08
CAS Number:7727-54-0
Formula:(NH₄)₂S₂O₈
MDL Number:MFCD00003390