


Ammonium peroxodisulphate (APS), EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:7727-54-0
:
Formula:(NH₄)₂S₂O₈
MDL Number:MFCD00003390
:228.2 g/mol
Merck Index:13,00545
Melting Pt:120 °C
Density:1.98 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-786-5
ADR:5.1,III
:M-Clarity™ quality level = MQ300
UN:1444
:GHS03
:GHS08
:GHS07
Specification Test Results
- Solubility:620 g/l (20 °C)
- Melting Point:120 °C (decomposition)
- Molar Mass:228.19 g/mol
- Bulk Density:900 kg/m3
- Density:1.98 g/cm3 (20 °C)
- pH:3.2 (100 g/l, H2O, 20 °C)
- Hazard Class:5.1
- Hazard Class Description:Oxidizer
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC1.01201.0500_TDS_EN ProductPage_101201
Product specification
msds_101201
SDS
Frequently Bought Together