


Ammonium peroxodisulphate (APS) 99+% for molecular biology DNase-, RNase-, protease-free
:7727-54-0
:
:(NH₄)₂S₂O₈
:MFCD00003390
:228.2 g/mol
:13,00545
:120 °C (dec.)
:1.98 g/cm³
:231-786-5
:1444
:5.1,III
:GHS07
:GHS08
:GHS03
:Ambient
- Appearance:White Powder
- Titration Iodometric:≥99 %
- Acidity:≤0.2 % (H2SO4)
- Dnase,Rnase,Protease activity:none detected
- Chloride (Cl):≤20 ppm
- Iron (Fe):≤10 ppm
- Heavy metals (as Pb):≤50 ppm
Frequently Bought Together