Ammonium dihydrogen phosphate, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Certificates
About this item
CAS Number:7722-76-1
:
Formula:NH₄H₂PO₄
MDL Number:MFCD00003396
Merck Index:13,00547
Melting Pt:190 °C
Density:1.8 g/cm³ (20 °C)
:115.03 g/mol
Storage Temperature:Ambient
EINECS:231-764-5
Related Information
Frequently Bought Together