Ammonium nitrate
Catalog # ROTHX988.3
Supplier: Roth Carl
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:2,5 kg
- Seq:4076
Specifications
About this item
:
:80.04 g/mol
Merck Index:13,00538
Boiling Pt:230 °C (1013 hPa)
Melting Pt:169 °C
Density:1.73 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:229-347-8
UN:1942
ADR:5.1,III
:GHS03
CAS Number:6484-52-2
Formula:NH₄NO₃
MDL Number:MFCD00011425