Ammonium bromide ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:12124-97-9
:
Formula:H₄BrN
MDL Number:MFCD00011419
:97.94 g/mol
Merck Index:14,505
Melting Pt:452 °C (subl.)
Density:2.429
EINECS:235-183-8
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together