Order Entry
Denmark
ContactUsLinkComponent
 
(Ethyl)triphenylphosphonium bromide 98%
 
undefined
(Ethyl)triphenylphosphonium bromide 98%

 

  • Environmentally Preferable:

 

 

  • Appearance:
    White to off-white Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    206.5 to 208.5 °C
  • Titration Argentometric:
    ≥97.5 % (on dry substance)
  • Loss on drying:
    ≤1 %

 

 

Frequently Bought Together