
Antimony(III) oxide, Supelco®
: Merck
:1309-64-4
:
:O₃Sb₂
:MFCD00011214
:291.52 g/mol
:13,00717
:1425 °C (1013 hPa)
:654 °C
:5.2 g/cm³ (20 °C)
:1549
:6.1,III
:M-Clarity™ quality level = MQ200
:215-175-0
:GHS08
:Ambient
- Solubility:2.70 mg/l (20 °C)
- Melting Point:656 °C (sublimed)
- Molar Mass:291.49 g/mol
- Bulk Density:800 - 1300 kg/m3
- Boiling Point:1550 °C (1013 hPa)
- Vapor Pressure:13.3 hPa (660 °C)
- Density:5.4 g/cm3
Frequently Bought Together