Tin(IV) oxide, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:18282-10-5
:
Formula:O₂Sn
MDL Number:MFCD00011244
:150.71 g/mol
Merck Index:14,08778
Boiling Pt:∼2500 °C (1013 hPa)
Melting Pt:1624.85 °C
Density:6.95 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:242-159-0
Specification Test Results
- Solubility:(20 °C) practically insoluble
- Melting Point:1630 °C
- Molar Mass:150.69 g/mol
- Bulk Density:500 - 600 kg/m3
- Boiling Point:1800 - 1900 °C (sublimed)
- Density:6.95 g/cm3 (20 °C)
- pH:4 - 5 (50 g/l, H2O, 20 °C) (slurry)
Specification Test Results
Related Information
Frequently Bought Together