Tin(IV) oxide 99.9% (trace metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:18282-10-5
:
Formula:O₂Sn
MDL Number:MFCD00011244
:150.71 g/mol
Merck Index:14,08778
Boiling Pt:∼2500 °C (1013 hPa)
Melting Pt:1624.85 °C
Density:6.95 g/cm³
EINECS:242-159-0
Specification Test Results
- Appearance:White to light grey Powder
- Total trace metal impurities:≤0.1 %
Specification Test Results
Frequently Bought Together