


Bly(II)oxid, gul ≥99.0% (by complexometric analysis), Puriss. p.a., Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:1317-36-8
:
Formula:PbO
MDL Number:MFCD00011164
:223.2 g/mol
Merck Index:13,05431
Boiling Pt:1,470 °C (1.013 hPa)
Melting Pt:890 °C
Density:9,66 g/cm³ (20 °C)
Storage Temperature:Stue temperatur
EINECS:215-267-0
UN:2291
ADR:6.1,III
:GHS07
:GHS08
:GHS09
Specification Test Results
- Assay (complexometric):99.0 - 101.0 %
- Calcium (Ca):Max. 10 ppm
- Cadmium (Cd):Max. 5 ppm
- Cobalt (Co):Max. 5 ppm
- Chromium (Cr):Max. 5 ppm
- Copper (Cu):Max. 5 ppm
- Iron (Fe):Max. 10 ppm
- Potassium (K):Max. 50 ppm
- Magnesium (Mg):Max. 5 ppm
- Manganese (Mn):Max. 5 ppm
- Sodium (Na):Max. 50 ppm
- Nickel (Ni):Max. 5 ppm
- Zinc (Zn):Max. 5 ppm
- Chloride (Cl):Max. 20 ppm
- Total N:Max. 10 ppm
Specification Test Results
Frequently Bought Together