Lead(II) carbonate basic ACS
Glass bottle for solids, 250 g
Supplier: Thermo Fisher Scientific
Lead(II) carbonate basic ACS
Catalog # 33328.30
Supplier: Thermo Fisher Scientific
CAS Number: 1319-46-6
Jump to:
Product Details & Documents
CAS No.: 1319-46-6 Molecular Formula: C2H2O8Pb3 Formula Weight: 775.63 Physical Form: Powder Density: 6.14 Appearance: White Odour: Odourless Melting Point: 400° dec. Solubility: Soluble in acids, insoluble in water or alcohol Merck Reference: 14,1005 UN No.: 3077 MDL No.: MFCD00078155
Ceramic glaze, exterior paint
Synonyms: Lead carbonat basic, White lead, Lead (II) carbonate hydroxide, Lead (II) hydroxide carbonate
- CAS Number:1319-46-6
- Formula:(PbCO₃)₂·Pb(OH)₂
- MDL Number:MFCD00078155
- Molecular Weight:775.63 g/mol
- Merck Index:14,1005
- Melting Pt:400 °C (dec.)
- Density:6.14
- Storage Temperature:Ambient
- EINECS:215-290-6
- UN:3077
- ADR:9,III
- :Density: 6.06 g/cm³ (20 °C)
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:250 g



