Lead(II) oxide low silver
About this item
CAS No.: 1317-36-8 Molecular Formula: PbO Formula Weight: 223.19 Physical Form: Powder Density: 9.53 Odour: Odourless Merck Reference: 14,5413 UN No.: 3077 MDL No.: MFCD00011164
As storage batteries, ceramic cements and fluxes, pottery and glazes, in chromium pigments, in oil refining, in paints, and in the assay of precious metal ores
- CAS Number:1317-36-8
- Formula:PbO
- MDL Number:MFCD00011164
- Molecular Weight:223.2 g/mol
- Merck Index:14,5413
- Boiling Pt:1470 °C (1013 hPa)
- Melting Pt:886 °C
- Density:9.53
- Storage Temperature:Ambient
- EINECS:215-267-0
- UN:3077
- ADR:9,III
2 Options Available Below
Product Details & Documents
CAS No.: 1317-36-8 Molecular Formula: PbO Formula Weight: 223.19 Physical Form: Powder Density: 9.53 Odour: Odourless Merck Reference: 14,5413 UN No.: 3077 MDL No.: MFCD00011164
As storage batteries, ceramic cements and fluxes, pottery and glazes, in chromium pigments, in oil refining, in paints, and in the assay of precious metal ores
- CAS Number:1317-36-8
- Formula:PbO
- MDL Number:MFCD00011164
- Molecular Weight:223.2 g/mol
- Merck Index:14,5413
- Boiling Pt:1470 °C (1013 hPa)
- Melting Pt:886 °C
- Density:9.53
- Storage Temperature:Ambient
- EINECS:215-267-0
- UN:3077
- ADR:9,III
Documents
Product Family Options
Product Information
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



