Aluminium nitride (≥32.0% N)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:24304-00-5
:
Formula:AlN
MDL Number:MFCD00003429
:40.99 g/mol
Merck Index:14,353
Boiling Pt:2517 °C (1013 hPa)
Melting Pt:>2200 °C
Density:3.26
EINECS:246-140-8
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together