Order Entry
Denmark
ContactUsLinkComponent
Calcium hydroxide ≥96%, GPR RECTAPUR®
Corrosive
Irritant
Calcium hydroxide ≥96%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Corrosive
Irritant
Calcium hydroxide ≥96%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay (at filling):
    Min. 96 %
  • Heavy metals (as Pb):
    Max. 50 ppm
  • Insolubility in hydrochloric acid:
    Max. 0.1 %
  • Not precipitated by (COONH4)2 (as SO4):
    Max. 2.5 %
  • Cl (Chloride):
    Max. 100 ppm
  • CO3 (as CaCO3):
    Max. 3 %
  • SO4 (Sulphate):
    Max. 0.2 %
  • Fe (Iron):
    Max. 0.05 %

Specification Test Results

Frequently Bought Together