Order Entry
Denmark
ContactUsLinkComponent
Magnesium oxide heavy, TECHNICAL
Magnesium oxide heavy, TECHNICAL
Catalog # 25056.460
Supplier:  VWR Chemicals
CAS Number:  
undefined
Magnesium oxide heavy, TECHNICAL
Catalog # 25056.460
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bucket
  • Pk:
    25 kg
  • Seq:
    4283

Specifications

Specification Test Results

  • Assay (on ignited basis):
    Min. 85%
  • Loss on ignition (800°C):
    Max. 8%
  • Silica:
    3 - 6 %

Specification Test Results

About this item

 :
 :40.3 g/mol
Merck Index:12,05713
Boiling Pt:3600 °C (1013 hPa)
Melting Pt:2800 °C
Density:3.58 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-171-9
CAS Number:1309-48-4
Formula:MgO
MDL Number:MFCD00011109