Order Entry
Denmark
ContactUsLinkComponent
 
Magnesium oxide light ≥99%, tested according to Ph. Eur., Fluka™
  63096-500G
 :  
Magnesium oxide light ≥99%, tested according to Ph. Eur., Fluka™
  63096-500G
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    500 g
  • Seq:
    3645

 

 

  • Identity B:
    complying
  • Assay (calculated to ignited substance):
    98.0 - 100.5 %
  • Appearance of the solution:
    complying
  • Loss on ignition:
    Max. 8.0 %
  • In water soluble matter:
    Max. 2.0 %
  • Insoluble in acetic acid:
    Max. 0.1 %
  • Arsenic (As):
    Max. 4 ppm
  • Calcium (Ca):
    Max. 1.5 %
  • Iron (Fe):
    Max. 0.1 %
  • Heavy metals (as Pb):
    Max. 30 ppm
  • Chloride (Cl):
    Max. 0.15 %
  • Sulfate (SO₄):
    Max. 1.0 %
  • Bulk Density:
    Max. 0.15 g/ml
  • Residual solvents:
    complying

 

 

 :
 :40.3 g/mol
 :3.600 °C (1.013 hPa)
 :2.852 °C
 :3.58 g/cm³ (20 °C)
 :Ambient
 :215-171-9
 :1309-48-4
 :MgO
 :MFCD00011109