Magnesium oxide
Catalog # ROTH8280.1
Supplier: Roth Carl
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
Specifications
About this item
:
:40.3 g/mol
Merck Index:13,05700
Boiling Pt:3600 °C (1013 hPa)
Melting Pt:2800 °C
Density:3.58 g/cm³ (25 °C)
EINECS:215-171-9
CAS Number:1309-48-4
Formula:MgO
MDL Number:MFCD00011109