Magnesium oxide light 98.0-100.5%, AnalaR NORMAPUR® analytical reagent
:1309-48-4
:
:MgO
:MFCD00011109
:40.3 g/mol
:3600 °C (1013 hPa)
:2800 °C
:3.58 g/cm³ (25 °C)
:215-171-9
- Assay (MgO)(calc. on ignited basis):98.0 - 100.5 %
- Insolubility in hydrochloric acid:Max. 0.02 %
- Loss on ignition (1000°C):Max. 4 %
- Solubility in water:Max. 0.7 %
- Total N (Nitrogen):Max. 20 ppm
- Cl (Chloride):Max. 0.05 %
- CO3 (as CO2):Max. 1.5 %
- SO4 (Sulphate):Max. 0.1 %
- Ca (Calcium):Max. 1.1 %
- Cu (Copper):Max. 10 ppm
- Fe (Iron):Max. 0.05 %
- K (Potassium):Max. 50 ppm
- Na (Sodium):Max. 3 %
- Pb (Lead):Max. 10 ppm
- Zn (Zinc):Max. 5 ppm
Frequently Bought Together