Magnesium oxide ≥99% (metals basis), nanopowder
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1309-48-4
:
Formula:MgO
MDL Number:MFCD00011109
:40.3 g/mol
Merck Index:14,5677
Boiling Pt:3600 °C
Melting Pt:2852 °C
Density:3.58
Storage Temperature:Ambient
EINECS:215-171-9
Related Information
Frequently Bought Together