Order Entry
Denmark
ContactUsLinkComponent
 
Calcium carbonate, EMSURE® Reag. Ph. Eur. for analysis, Supelco®
 :  Merck
 
Calcium carbonate, EMSURE® Reag. Ph. Eur. for analysis, Supelco®
 :  Merck
 

 

  • Environmentally Preferable:

 

 

  • Solubility:
    0.014 g/l (20 °C)
  • Melting Point:
    825 °C (decomposition)
  • Molar Mass:
    100.09 g/mol
  • Bulk Density:
    400 - 1400 kg/m3
  • pH:
    9.5 - 10.5 (100 g/l, H2O, 20 °C) (slurry)

 

Frequently Bought Together