Barium bromide, ultra dry ≥99.998% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10553-31-8
:
Formula:BaBr₂
MDL Number:MFCD00003444
:297.14 g/mol
Merck Index:14,968
Boiling Pt:1835 °C (1013 hPa)
Melting Pt:850 °C
Density:4.781
UN:1564
ADR:6.1,III
:ampouled under argon
EINECS:234-140-0
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together