Barium bromide, anhydrous 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10553-31-8
:
Formula:BaBr₂
MDL Number:MFCD00003444
Boiling Pt:1835
Melting Pt:850 °C
Density:4.781
Molecular Weight:297.14 g/mol
EINECS:234-140-0
ADR:6.1 III
Merck Index:14,968
UN:1564
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together