Order Entry
Denmark
ContactUsLinkComponent
Potassium sulphate, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Potassium sulphate, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Supplier:  Merck
Certificates
Potassium sulphate, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    111 g/l (20 °C)
  • Melting Point:
    1069 °C
  • Molar Mass:
    174.26 g/mol
  • Bulk Density:
    800 kg/m3
  • Boiling Point:
    1689 °C (1013 hPa)
  • Density:
    2.66 g/cm3 (20 °C)
  • pH:
    5.5 - 8.5 (50 g/l, H2O, 20 °C)

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together