Potassium thiocyanate, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:333-20-0
:
Formula:KSCN
MDL Number:MFCD00011413
Merck Index:12,07866
Boiling Pt:500 °C (1013 hPa)
Melting Pt:175 °C
Density:1.61 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ300
:97.18 g/mol
Storage Temperature:Ambient
EINECS:206-370-1
:GHS05
:GHS07
Specification Test Results
- Solubility:208 g/l (20 °C)
- Melting Point:175 °C
- Molar Mass:97.18 g/mol
- Bulk Density:750 - 1000 kg/m3
- Vapor Pressure:- 1 hPa (20 °C)
- Density:1.89 g/cm3 (20 °C)
- pH:5.3 - 8.5 (50 g/l, H2O, 20 °C)
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC1.05125.0250_TDS_EN ProductPage_105125
Product specification
msds_105125
SDS
Frequently Bought Together