Potassium thiocyanate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Potassium rhodanide
CAS Number:333-20-0
:
Formula:KSCN
MDL Number:MFCD00011413
Merck Index:14,7691
Boiling Pt:500 °C (dec.)
Melting Pt:172…175 °C
Density:1.89
:97.18 g/mol
EINECS:206-370-1
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together