di-Potassium hydrogen phosphate
Glass bottle for solids, 250 g
Catalog # SERA39767.01
Marketsource
Supplier: Serva
di-Potassium hydrogen phosphate
Catalog # SERA39767.01
Supplier: Serva
Marketsource
CAS Number: 7758-11-4
Jump to:
Product Details & Documents
Synonyms: di-Potassium hydrogen orthophosphate, Dipotassium hydrogenphosphate, Potassium hydrogen phosphate, Dipotassium hydrogen phosphate, Dipotassium Phosphate, Dibasic Potassium Phosphate, Potassium Phosphate, Dibasic, (dipotassium hydrogen phosphate), Potassium phosphate dibasic, Dipotassium phosphate
- CAS Number:7758-11-4
- Formula:K₂HPO₄
- MDL Number:MFCD00011383
- Molecular Weight:174.18 g/mol
- Merck Index:13,07743
- Melting Pt:340 °C
- Density:2.3 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-834-5
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:250 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



