≥99.0%, powder, Ultrapure Bioreagent for liquid chromatography, for molecular biology, J.T.Baker®
Supplier: Avantor
Certificates
About this item
Synonyms: di-Potassium hydrogen orthophosphate, Dipotassium hydrogenphosphate, Potassium hydrogen phosphate, Dipotassium hydrogen phosphate, Dipotassium Phosphate, Dibasic Potassium Phosphate, Potassium Phosphate, Dibasic, (dipotassium hydrogen phosphate), Potassium phosphate dibasic, Dipotassium phosphate
CAS Number:7758-11-4
:
Formula:K₂HPO₄
MDL Number:MFCD00011383
Density:2.3 g/cm³ (20 °C)
Molecular Weight:174.18 g/mol
Melting Pt:340
Merck Index:13,07743
Storage Temperature:Ambient
EINECS:231-834-5
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:= 99.0 %
- Assay (K₂HPO₄):≥99.0 %
- DNase Activity (None Detected):Passes Test
- RNase Activity:Passes Test
- Protease Activity:Passes Test
- pH of 5% Solution at 25°C:8.5 - 9.6
- Chloride (Cl):≤0.001 %
- Insoluble Matter:≤0.01 %
- Trace Impurities - Iron (Fe):≤0.001 %
- Loss on Drying at 105°C:≤1.0 %
- Trace Impurities - ACS - Heavy Metals (as Pb):≤5 ppm
Specification Test Results
Frequently Bought Together