Potassium iodate ≥99.5% ACS, Fluka™
Catalog # 215929-100G
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 g
- Seq:3401
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:99.4 - 100.4 %
- pH (5 %, 25 °C):5.0 - 8.0
- Water insoluble matter:Max. 0.005 %
- Iron (Fe):Max. 0.001 %
- Sodium (Na):Max. 0.005 %
- Heavy metals (as Pb):Max. 5 ppm
- Bromide, chloride (as Cl):Max. 0.01 %
- Iodide (I):Max. 0.001 %
- Sulfate (SO₄):Max. 0.005 %
- Total N:Max. 0.005 %
Specification Test Results
About this item
:
:214 g/mol
EINECS:231-831-9
UN:1479
:GHS03
:GHS07
Storage Temperature:Ambient
CAS Number:7758-05-6
Formula:KlO₃
MDL Number:MFCD00011406
Melting Pt:560 °C
Density:3.93 g/cm³ (20 °C)
ADR:5.1 II
Merck Index:13,07726