Order Entry
Denmark
ContactUsLinkComponent
 
Sulphuric acid ≥96%, VLSI Selectipur® for the electronics industry
  57384943.
 :  
undefined
Sulphuric acid ≥96%, VLSI Selectipur® for the electronics industry
  57384943.
 :  

 

  • Pack type:
    Plastic drum
  • Pk:
    340 kg
  • Seq:
    4811

 

 

 :
 :98.08 g/mol
 :14,08974
 :330 °C (1013 hPa)
 :10.38 °C
 :1.84 g/cm³ (20 °C)
 :Ambient
 :231-639-5
 :1830
 :8,II
 :01-2119458838-20
 :GHS05
 :7664-93-9
 :H₂SO₄
 :MFCD00064589