Water, CHROMASOLV™ for headspace gas chromatography, Riedel-de Haën™
Supplier: Honeywell Chemicals
Certificates
About this item
Synonyms: Hydrogen oxide
CAS Number:7732-18-5
:
Formula:H₂O
MDL Number:MFCD00011332
:18.02 g/mol
Merck Index:13,10098
Boiling Pt:100 °C
Melting Pt:0 °C
Density:1 g/cm3
Storage Temperature:Ambient
EINECS:231-791-2
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Refractive Index:n20/D 1.34 (lit.)
- Suitability for the GC Headspace:complying
Specification Test Results
Frequently Bought Together