1-Amino-9,10-anthraquinone 97%, Sigma-Aldrich®
Catalog # A39009-100G
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
Specification Test Results
- Appearance (Form):Powder or Crystal or Chunk(s)
- Infrared spectrum:Conforms to Structure
- Carbon:72.7 - 78.0 %
- Nitrogen:6.0 - 6.6 %
- Purity (HPLC):≥96.5 %
Specification Test Results
About this item
:
:223.23 g/mol
Merck Index:13,00416
Boiling Pt:465 °C (1013 hPa)
Melting Pt:251…254 °C
:M-Clarity™ quality level = MQ100
Storage Temperature:Room Temperature
EINECS:201-423-5
CAS Number:82-45-1
Formula:C₁₄H₉NO₂
MDL Number:MFCD00001213