1-Amino-9,10-anthraquinone for synthesis, Sigma-Aldrich®
Catalog # 8.41590.0100
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 g
- Seq:3401
Specifications
Specification Test Results
- Melting Point:251 - 254 °C
- Molar Mass:223.23 g/mol
- Hill Formula:C14H9NO2
Specification Test Results
About this item
:
:223.23 g/mol
Merck Index:13,00416
Boiling Pt:465 °C (1013 hPa)
Melting Pt:251…254 °C
:M-Clarity™ quality level = MQ200
EINECS:201-423-5
CAS Number:82-45-1
Formula:C₁₄H₉NO₂
MDL Number:MFCD00001213